| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047437.hg.1 |
TC01003042.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047437.hg.1 |
ENST00000207157.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047437.hg.1 |
ENST00000344218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047437.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047433.hg.1 |
TC01003042.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047433.hg.1 |
ENST00000207157.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047433.hg.1 |
ENST00000344218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047433.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047433.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047434.hg.1 |
TC01003042.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047434.hg.1 |
ENST00000207157.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047434.hg.1 |
ENST00000344218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047434.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047434.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047432.hg.1 |
TC01003042.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047432.hg.1 |
ENST00000207157.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047432.hg.1 |
ENST00000344218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047432.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047432.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215416.hg.1 |
TC0100015445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215419.hg.1 |
TC0100015445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215418.hg.1 |
TC0100015445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100215422.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215422.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215422.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215422.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215422.hg.1 |
TC0100015445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
ENST00000207157.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
ENST00000369429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
NM_152380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215417.hg.1 |
TC0100015445.hg.1 |
Clariom D TC |