| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000440012.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000440012.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000440012.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02002768.hg.1 |
ENST00000379558.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02002768.hg.1 |
ENST00000403861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02002768.hg.1 |
ENST00000465300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02002768.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02002768.hg.1 |
TC02000191.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200149064.hg.1 |
TC0200007113.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000379558.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000401723.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000420297.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000440012.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000440012.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000440012.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000460951.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000465300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
ENST00000475900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007113.hg.1 |
NM_199280.2 |
Refseq |