Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033054.2 | NP_149043.2 | 20 - 21 | 2893 | 157 | |
XM_017012503.1 | XP_016867992.1 | 19 - 20 | 2652 | 157 | ||
XM_017012504.1 | XP_016867993.1 | 20 - 21 | 2888 | 157 | ||
GenBank mRNA | AA651942.1 | - | 2 - 3 | 156 | ||
AB290179.1 | - | 20 - 21 | 2853 | 157 | ||
AK131089.1 | - | 15 - 16 | 5010 | 157 | ||
AL832302.1 | - | 19 - 20 | 2648 | 157 | ||
BC015693.2 | - | 4 - 5 | 583 | 157 | ||
BC113544.1 | - | 20 - 21 | 2840 | 8322596 | 157 | |
BC117241.1 | - | 20 - 21 | 2840 | 40125792 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |