Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01075903_m1
          See other MIB1 GE Assays ›
          Gene Symbol:
          MIB1
          Gene Name:
          MIB E3 ubiquitin protein ligase 1
          Gene Aliases:
          DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6
          Chromosome Location:
          Chr.18: 21704916 - 21870953 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01075903_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020774.4 NP_065825.1 5 - 6 1493 88
          XM_017025874.2 XP_016881363.1 5 - 6 1493 88
          XM_017025875.2 XP_016881364.1 5 - 6 1493 88
          XM_047437676.1 XP_047293632.1 5 - 6 680 88
          GenBank mRNA AK091266.1 - 4 - 5 386 88
          AY147849.1 - 5 - 6 974 88
          AY149908.1 - 5 - 6 701 88
          BC110581.1 - 5 - 6 701 88
          BC110582.1 - 5 - 6 701 88
          CR749635.1 - 2 - 3 121 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800006841.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MIB1

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          in utero embryonic development
          somitogenesis
          neural tube formation
          heart looping
          ubiquitin-dependent protein catabolic process
          endocytosis
          Notch signaling pathway
          heart development
          protein ubiquitination
          central nervous system neuron differentiation
          negative regulation of neuron differentiation
          positive regulation of endocytosis
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          transferase activity
          metal ion binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 ENST00000582102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000577749.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578260.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 ENST00000582102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay TC1800006841.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 ENST00000261537.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 ENST00000578646.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 NM_020774.3 Refseq
          Affymetrix Human Clariom D Assay PSR1800146860.hg.1 TC1800006841.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0