Search
Search

CPLX3| Gene Symbol: CPLX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030005.3 | NP_001025176.1 | 1 - 2 | 244 | 70 | |
| GenBank mRNA | AB072900.1 | - | 1 - 2 | 244 | 70 | |
| AK024055.1 | - | 1 - 2 | 261 | 70 | ||
| AY286501.1 | - | 1 - 2 | 181 | 70 | ||
| BC018026.1 | - | 1 - 2 | 244 | 70 | ||
| GQ900997.1 | - | 1 - 2 | 181 | 70 | ||
Set Membership: |
|