Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01081249_m1
          See other SLC25A24 GE Assays ›
          Gene Symbol:
          SLC25A24
          Gene Name:
          solute carrier family 25 member 24
          Gene Aliases:
          APC1, SCAMC-1
          Chromosome Location:
          Chr.1: 108134722 - 108200358 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01081249_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC25A24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013386.4 NP_037518.3 9 - 10 1468 84
          NM_213651.2 NP_998816.1 9 - 10 1379 84
          XM_017001104.1 XP_016856593.1 5 - 6 724 84
          GenBank mRNA AJ619961.1 - 9 - 10 1311 84
          AJ619987.1 - 9 - 10 1257 84
          AK292567.1 - 9 - 10 1379 84
          AK304165.1 - 9 - 10 1462 84
          AK316304.1 - 9 - 10 1422 84
          AK316506.1 - 9 - 10 1290 84
          BC014519.1 - 9 - 10 1324 3842039 84
          BC068561.1 - 9 - 10 1357 5259336 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015160.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002942.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC25A24

          Panther Classification:

          Molecular Function -

          mitochondrial carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          mitochondrial transport
          regulation of cell death
          ATP transport
          cellular response to oxidative stress
          transmembrane transport
          cellular response to calcium ion
          structural constituent of ribosome
          ATP transmembrane transporter activity
          calcium ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01045710.hg.1 TC01002942.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01045710.hg.1 ENST00000264128.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01045710.hg.1 ENST00000370041.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01045710.hg.1 NM_013386.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01045710.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100212924.hg.1 TC0100015160.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 ENST00000569674.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay TC0100015160.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 ENST00000264128.13 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 ENST00000370041.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 ENST00000565488.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 NM_013386.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 NM_213651.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100212925.hg.1 TC0100015160.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 214599 - -
          ABI Human Genome Survey Microarray Version 2.0 219882 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline