| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013723.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013723.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013723.hg.1 |
TC22000808.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000490326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000490326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
ENST00000490326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163554.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163554.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163554.hg.1 |
TC2200008759.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200163554.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163554.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000325157.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000490326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000490326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
ENST00000490326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008759.hg.1 |
NM_152512.3 |
Refseq |