| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003961.hg.1 |
ENST00000373433.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003961.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003961.hg.1 |
TC20000278.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003962.hg.1 |
ENST00000373433.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003962.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003962.hg.1 |
TC20000278.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000471511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000484683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000614670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000618318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
ENST00000622494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007300.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
ENST00000495457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150475.hg.1 |
TC2000007300.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
ENST00000373433.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
ENST00000449186.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
ENST00000462548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
NM_021215.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150476.hg.1 |
TC2000007300.hg.1 |
Clariom D TC |