| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016611.hg.1 |
ENST00000295645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016611.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016611.hg.1 |
TC04001215.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016612.hg.1 |
ENST00000295645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016612.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016612.hg.1 |
TC04001215.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400173210.hg.1 |
TC0400010749.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010749.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
ENST00000295645.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
ENST00000615028.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
NM_152401.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400173209.hg.1 |
TC0400010749.hg.1 |
Clariom D TC |