Search
Search

FREM3| Gene Symbol: FREM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168235.1 | NP_001161707.1 | 1 - 2 | 5191 | 75 | |
| GenBank mRNA | AW953229.1 | - | 1 - 2 | 334 | 75 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Oligo Microarray 4x44K | 24410 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26682 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P14932275 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P36945363 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01096550 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16888820 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01096565 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16888960 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16889067 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16888909 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P36945331 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16888835 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P36945331 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16888820 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16888909 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P738168 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493958 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493966 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654589 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654556 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493935 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654603 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493965 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493930 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493951 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493963 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493951 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493966 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654603 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493965 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493963 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654589 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654556 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493958 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493935 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493930 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15621651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23654604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03493939 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8102986 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 243337_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022366.hg.1 | TC04001604.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022366.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022366.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022365.hg.1 | TC04001604.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022365.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022365.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000508899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | ENST00000508899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182403.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182403.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182403.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182403.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182403.hg.1 | TC0400012011.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000508899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | ENST00000508899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012011.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008884.hg.1 | ENST00000499587.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182404.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182404.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182404.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182404.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182404.hg.1 | TC0400012011.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400182405.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182405.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182405.hg.1 | ENST00000329798.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182405.hg.1 | NM_001168235.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182405.hg.1 | TC0400012011.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 243337_at | - | - |
| Human Genome U133 Plus 2.0 Array | 243337_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243337_at5 | - | - |
| Human Genome U95 Set | 62731_at | - | - |
| Human Genome U95 Set | 62731_PM_at | - | - |
| Human Genome U133 Set | 243337_at | - | - |
| Human Genome U133 Set | 243337_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024801 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024801 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 131424 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 306621 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1686928 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1686928 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1686928 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1686928 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1686928 | - | - |