Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01085821_g1
          See other GATA1 GE Assays ›
          Gene Symbol:
          GATA1
          Gene Name:
          GATA binding protein 1
          Gene Aliases:
          ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT
          Chromosome Location:
          Chr.X: 48786540 - 48794311 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01085821_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GATA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002049.3 NP_002040.1 2 - 3 308 80
          XM_011543897.2 XP_011542199.1 2 - 3 342 80
          GenBank mRNA BC009797.1 - 2 - 3 311 4048082 80
          M30601.1 - 2 - 3 275 80
          X17254.1 - 2 - 3 329 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00007204.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000260.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GATA1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          transcription from RNA polymerase II promoter
          cell-cell signaling
          heart development
          blood coagulation
          negative regulation of cell proliferation
          male gonad development
          organ morphogenesis
          tissue development
          regulation of glycoprotein biosynthetic process
          regulation of definitive erythrocyte differentiation
          regulation of primitive erythrocyte differentiation
          erythrocyte differentiation
          megakaryocyte differentiation
          platelet formation
          basophil differentiation
          eosinophil differentiation
          negative regulation of bone mineralization
          positive regulation of osteoblast proliferation
          embryonic hemopoiesis
          eosinophil fate commitment
          negative regulation of apoptotic process
          positive regulation of erythrocyte differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          erythrocyte development
          homeostasis of number of cells within a tissue
          positive regulation of peptidyl-tyrosine phosphorylation
          platelet aggregation
          transcriptional activation by promoter-enhancer looping
          dendritic cell differentiation
          cellular response to thyroid hormone stimulus
          negative regulation of transcription regulatory region DNA binding
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          p53 binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          zinc ion binding
          DNA binding, bending
          chromatin DNA binding
          sequence-specific DNA binding
          C2H2 zinc finger domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X003230.hg.1 TC0X000260.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X003230.hg.1 ENST00000376665.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X003230.hg.1 ENST00000376670.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X003230.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00007204.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 ENST00000376665.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 ENST00000376670.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 NM_002049.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00149271.hg.1 TC0X00007204.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline