| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003233.hg.1 |
TC0X000260.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003233.hg.1 |
ENST00000376665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003233.hg.1 |
ENST00000376670.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003233.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003232.hg.1 |
TC0X000260.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003232.hg.1 |
ENST00000376665.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003232.hg.1 |
ENST00000376670.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X003232.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007204.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149277.hg.1 |
TC0X00007204.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149278.hg.1 |
TC0X00007204.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
ENST00000376665.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
ENST00000376670.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
NM_002049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00149273.hg.1 |
TC0X00007204.hg.1 |
Clariom D TC |