| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015320.hg.1 |
ENST00000311069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015320.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015320.hg.1 |
TC19001115.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009488.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
ENST00000311069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
ENST00000598935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
NM_174895.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176281.hg.1 |
TC1900009488.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176285.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176285.hg.1 |
NM_001271830.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176285.hg.1 |
TC1900009488.hg.1 |
Clariom D TC |