| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023909.hg.1 |
ENST00000301420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023909.hg.1 |
ENST00000448701.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023909.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023909.hg.1 |
TC19001758.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011223.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194314.hg.1 |
TC1900011223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000301420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000593325.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000593859.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
ENST00000596300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
NM_002257.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194312.hg.1 |
TC1900011223.hg.1 |
Clariom D TC |