Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01087603_m1
          See other PYGO1 GE Assays ›
          Gene Symbol:
          PYGO1 POLR2M
          Gene Name:
          pygopus family PHD finger 1
          RNA polymerase II subunit M
          Gene Aliases:
          -
          GCOM1, GRINL1A, Gdown, Gdown1
          Chromosome Location:
          Chr.15: 55538884 - 55588971 on Build GRCh38
          Chromosome Location:
          Chr.15: 57706714 - 57717557 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01087603_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PYGO1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001330326.2 NP_001317255.1 2 - 3 600 87
          NM_001367806.1 NP_001354735.1 2 - 3 600 87
          NM_015617.3 NP_056432.1 2 - 3 257 87
          XM_047432381.1 XP_047288337.1 2 - 3 246 87
          GenBank mRNA AF457207.1 - 2 - 3 138 87
          EF625686.1 - 3 - 4 263 87
          Gene Symbol: POLR2M
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY339380.1 - 3 - 4 263 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009504.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001468.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_047432381.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PYGO1
          Click here to find siRNAs for Gene POLR2M

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          hematopoietic progenitor cell differentiation
          chromatin organization
          spermatid development
          spermatid nucleus differentiation
          Wnt signaling pathway
          protein localization to nucleus
          positive regulation of transcription from RNA polymerase II promoter
          canonical Wnt signaling pathway
          protein binding
          zinc ion binding
          metal ion binding
          histone H3K4me3 reader activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15013750.hg.1 ENST00000302000.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15013750.hg.1 ENST00000401688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15013750.hg.1 NM_015617.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15013750.hg.1 TC15001468.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500170120.hg.1 TC1500009504.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000302000.10 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 ENST00000563719.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 NM_015617.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009504.hg.1 NM_015617.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline