| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006177.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006177.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006177.hg.1 |
TC20000456.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006176.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006176.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20006176.hg.1 |
TC20000456.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007887.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154724.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154724.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154724.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154724.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154724.hg.1 |
TC2000007887.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154725.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154725.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154725.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154725.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154725.hg.1 |
TC2000007887.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154726.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154726.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154726.hg.1 |
ENST00000371168.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154726.hg.1 |
NM_178456.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154726.hg.1 |
TC2000007887.hg.1 |
Clariom D TC |