| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019136.hg.1 |
TC09001533.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019136.hg.1 |
ENST00000265922.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019136.hg.1 |
ENST00000373964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019136.hg.1 |
ENST00000373969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019136.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000482797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
ENST00000482797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000482797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
ENST00000482797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011370.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
ENST00000265922.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
ENST00000373964.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
NM_014618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177389.hg.1 |
TC0900011370.hg.1 |
Clariom D TC |