| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012536.hg.1 |
ENST00000249373.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012536.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07012536.hg.1 |
TC07000795.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000475779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
ENST00000495998.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009082.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
ENST00000249373.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
ENST00000462420.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
NM_005631.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700164645.hg.1 |
TC0700009082.hg.1 |
Clariom D TC |