| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009807.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009807.hg.1 |
ENST00000432306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009807.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009807.hg.1 |
TC10000814.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000161079.hg.1 |
TC1000008927.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000393077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000432306.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000472892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000480057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000483122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000489357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000494728.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
ENST00000496445.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008927.hg.1 |
NM_145206.3 |
Refseq |