| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000467657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000467657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160700.hg.1 |
TC0200008680.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200160701.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160701.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160701.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160701.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160701.hg.1 |
TC0200008680.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000233969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000467657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000467657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
ENST00000469286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008680.hg.1 |
NM_003048.5 |
Refseq |