Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01091917_g1
          See other ZCRB1 GE Assays ›
          Gene Symbol:
          ZCRB1
          Gene Name:
          zinc finger CCHC-type and RNA binding motif containing 1
          Gene Aliases:
          MADP-1, MADP1, RBM36, SNRNP31, ZCCHC19
          Chromosome Location:
          Chr.12: 42312086 - 42326094 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01091917_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZCRB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033114.4 NP_149105.3 5 - 6 507 81
          XM_017020124.2 XP_016875613.1 4 - 5 421 81
          GenBank mRNA AB062361.1 - 5 - 6 543 81
          AK308244.1 - 5 - 6 413 81
          BC005099.1 - 5 - 6 493 81
          BC010177.2 - 5 - 6 508 81
          BC022543.1 - 5 - 6 518 81
          BI666919.1 - 1 - 2 82 81
          BK005200.1 - 4 - 5 334 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010454.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001403.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZCRB1

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          RNA splicing
          nucleic acid binding
          RNA binding
          protein binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000551102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000551102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 ENST00000610611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010454.hg.1 NR_107005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018164.hg.1 ENST00000266529.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12018164.hg.1 ENST00000552673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12018164.hg.1 NM_033114.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12018164.hg.1 TC12001403.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200178813.hg.1 TC1200010454.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000266529.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000548022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000551102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000551102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552235.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000552673.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 ENST00000610611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NM_033114.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010454.hg.1 NR_107005.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline