Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01092438_m1
          See other DHCR24 GE Assays ›
          Gene Symbol:
          DHCR24
          Gene Name:
          24-dehydrocholesterol reductase
          Gene Aliases:
          DCE, Nbla03646, SELADIN1, seladin-1
          Chromosome Location:
          Chr.1: 54849627 - 54887195 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01092438_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DHCR24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014762.4 NP_055577.1 6 - 7 77
          GenBank mRNA AB073391.1 - 7 - 8 1021 77
          AF261758.1 - 6 - 7 1116 77
          AK298414.1 - 1 - 2 375 77
          AK302774.1 - 6 - 7 977 77
          AK304437.1 - 6 - 7 863 77
          AK316419.1 - 6 - 7 908 77
          AK316427.1 - 6 - 7 847 77
          BC004375.1 - 6 - 7 77
          BC011669.2 - 6 - 7 1079 77
          D13643.3 - 6 - 7 1129 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014276.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002694.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DHCR24

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          steroid biosynthetic process
          cholesterol biosynthetic process
          Ras protein signal transduction
          protein localization
          steroid metabolic process
          cholesterol metabolic process
          negative regulation of cell proliferation
          response to hormone
          tissue development
          sterol metabolic process
          sterol biosynthetic process
          male genitalia development
          plasminogen activation
          cholesterol biosynthetic process via desmosterol
          cholesterol biosynthetic process via lathosterol
          amyloid precursor protein catabolic process
          skin development
          membrane organization
          delta24(24-1) sterol reductase activity
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-CH group of donors, NAD or NADP as acceptor
          enzyme binding
          peptide antigen binding
          delta24-sterol reductase activity
          flavin adenine dinucleotide binding
          FAD binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042250.hg.1 ENST00000371269.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042250.hg.1 ENST00000535035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042250.hg.1 ENST00000537443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042250.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042250.hg.1 TC01002694.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042251.hg.1 ENST00000371269.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042251.hg.1 ENST00000535035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042251.hg.1 ENST00000537443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042251.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042251.hg.1 TC01002694.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206989.hg.1 TC0100014276.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206990.hg.1 TC0100014276.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline