Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01092512_g1
          See other TIMP1 GE Assays ›
          Gene Symbol:
          TIMP1
          Gene Name:
          TIMP metallopeptidase inhibitor 1
          Gene Aliases:
          CLGI, EPA, EPO, HCI, TIMP, TIMP-1
          Chromosome Location:
          Chr.X: 47582291 - 47586791 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01092512_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TIMP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003254.2 NP_003245.1 3 - 4 394 78
          XM_017029766.1 XP_016885255.1 3 - 4 348 78
          GenBank mRNA A10416.1 - 3 - 4 264 78
          AK074854.1 - 3 - 4 372 78
          AK310206.1 - 3 - 4 246 78
          AK311937.1 - 3 - 4 246 78
          BC000866.1 - 3 - 4 347 3460153 78
          BC007097.1 - 3 - 4 295 78
          BQ181804.1 - 3 - 4 538 78
          BU857950.1 - 441 78
          CR407638.1 - 2 - 3 202 78
          CR541982.1 - 2 - 3 202 78
          M12670.1 - 3 - 4 250 78
          M59906.1 - 3 - 4 245 78
          S68252.1 - 2 - 3 205 78
          X02598.1 - 3 - 4 243 78
          X03124.1 - 3 - 4 264 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00007149.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000238.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017029766.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TIMP1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell activation
          platelet degranulation
          aging
          positive regulation of cell proliferation
          response to hormone
          negative regulation of endopeptidase activity
          extracellular matrix disassembly
          response to cytokine
          wound healing
          negative regulation of apoptotic process
          negative regulation of catalytic activity
          response to peptide hormone
          negative regulation of membrane protein ectodomain proteolysis
          cartilage development
          negative regulation of trophoblast cell migration
          regulation of integrin-mediated signaling pathway
          protease binding
          cytokine activity
          protein binding
          growth factor activity
          metalloendopeptidase inhibitor activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X002815.hg.1 ENST00000218388.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002815.hg.1 ENST00000377017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002815.hg.1 ENST00000377018.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002815.hg.1 ENST00000456754.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002815.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X002815.hg.1 TC0X000238.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X002814.hg.1 ENST00000218388.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002814.hg.1 ENST00000377017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002814.hg.1 ENST00000377018.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002814.hg.1 ENST00000456754.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X002814.hg.1 NM_003254.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X002814.hg.1 TC0X000238.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000445623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00007149.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00148753.hg.1 TC0X00007149.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000218388.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000377017.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000441738.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 ENST00000456754.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 NM_003254.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00148752.hg.1 TC0X00007149.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline