Search
Search

PITPNM2| Gene Symbol: PITPNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300801.2 | NP_001287730.1 | 17 - 18 | 3085 | 80 | |
| NM_001384668.1 | NP_001371597.1 | 17 - 18 | 3121 | 80 | ||
| XM_047429202.1 | XP_047285158.1 | 17 - 18 | 2976 | 80 | ||
| XM_047429203.1 | XP_047285159.1 | 17 - 18 | 2928 | 80 | ||
| XM_047429204.1 | XP_047285160.1 | 17 - 18 | 2925 | 80 | ||
| XM_047429207.1 | XP_047285163.1 | 17 - 18 | 2976 | 80 | ||
| XM_047429208.1 | XP_047285164.1 | 17 - 18 | 2958 | 80 | ||
| XM_047429211.1 | XP_047285167.1 | 17 - 18 | 2925 | 80 | ||
| GenBank mRNA | AB040890.2 | - | 17 - 18 | 2769 | 80 | |
Set Membership: |
|