Search
Search

SERPINA2| Gene Symbol: SERPINA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006220.3 | NP_006211.2 | 1216 | 70 | ||
| GenBank mRNA | DB526949.1 | - | 177 | 70 | ||
| JX680599.1 | - | 1073 | 70 | |||
Set Membership: |
|