Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01099594_m1
          See other BMP6 GE Assays ›
          Gene Symbol:
          BMP6
          Gene Name:
          bone morphogenetic protein 6
          Gene Aliases:
          IO, VGR, VGR1
          Chromosome Location:
          Chr.6: 7726099 - 7881728 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01099594_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BMP6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001718.6 NP_001709.1 1 - 2 1522 108
          GenBank mRNA AK300628.1 - 2 - 3 803 108
          BC096084.2 - 2 - 3 411 108
          BC096085.2 - 2 - 3 411 108
          BC099624.1 - 2 - 3 411 108
          BC099625.1 - 2 - 3 575 108
          BC111763.1 - 2 - 3 575 108
          BC112352.1 - 2 - 3 575 108
          BC113018.1 - 2 - 3 470 108
          BC113019.1 - 2 - 3 467 108
          BC128031.1 - 2 - 3 367 108
          M60315.1 - 1 - 2 824 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006873.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000062.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human Growth Factors HUMAN SMAD SIGNALING NETWORK Inventoried Human_Growth_Factors Human_SMAD_Signaling_Network Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BMP6

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          ossification
          osteoblast differentiation
          eye development
          kidney development
          positive regulation of endothelial cell proliferation
          endochondral ossification
          type B pancreatic cell development
          cellular iron ion homeostasis
          inflammatory response
          immune response
          heart development
          positive regulation of cell proliferation
          tissue development
          response to iron ion
          positive regulation of gene expression
          response to activity
          cell differentiation
          neuron differentiation
          positive regulation of bone mineralization
          BMP signaling pathway
          male genitalia development
          positive regulation of lipopolysaccharide-mediated signaling pathway
          response to magnesium ion
          positive regulation of chondrocyte differentiation
          positive regulation of aldosterone biosynthetic process
          response to retinoic acid
          regulation of cell proliferation
          positive regulation of vascular permeability
          positive regulation of endothelial cell differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cell development
          positive regulation of epithelial cell proliferation
          positive regulation of protein secretion
          positive regulation of developmental process
          cartilage development
          response to glucocorticoid
          bone development
          positive regulation of SMAD protein import into nucleus
          multicellular organismal iron ion homeostasis
          cellular response to mechanical stimulus
          cellular response to iron ion
          cellular response to BMP stimulus
          positive regulation of intracellular signal transduction
          negative regulation of adherens junction organization
          negative regulation of cell-cell adhesion mediated by cadherin
          positive regulation of aldosterone secretion
          cytokine activity
          protein binding
          growth factor activity
          protein heterodimerization activity
          BMP receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000603.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000603.hg.1 ENST00000540959.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000603.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000603.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000599.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000599.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000599.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000604.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000604.hg.1 ENST00000537240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000604.hg.1 ENST00000540959.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000604.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000604.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000602.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000602.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000602.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000605.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000605.hg.1 ENST00000537240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000605.hg.1 ENST00000540959.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000605.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000605.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000601.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000601.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000601.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600147080.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147080.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147080.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147080.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147080.hg.1 TC0600006873.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600147078.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147078.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147078.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147078.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147078.hg.1 TC0600006873.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147079.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147079.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147079.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147079.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147079.hg.1 TC0600006873.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline