| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000607.hg.1 |
TC06000062.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000607.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000607.hg.1 |
ENST00000537240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000607.hg.1 |
ENST00000540959.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000607.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147082.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147082.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147082.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147082.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147082.hg.1 |
TC0600006873.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
ENST00000283147.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006873.hg.1 |
NM_001718.5 |
Refseq |