Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01099598_gH
          See other BMP6 GE Assays ›
          Gene Symbol:
          BMP6
          Gene Name:
          bone morphogenetic protein 6
          Gene Aliases:
          VGR, VGR1
          Chromosome Location:
          Chr.6: 7726778 - 7881728 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01099598_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BMP6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001718.4 NP_001709.1 5 - 6 1460 64
          GenBank mRNA AK300628.1 - 6 - 7 1420 64
          BC096084.2 - 6 - 7 1028 64
          BC096085.2 - 6 - 7 1028 64
          BC099624.1 - 6 - 7 1028 64
          BC099625.1 - 6 - 7 1192 64
          BC111763.1 - 6 - 7 1192 64
          BC112352.1 - 6 - 7 1192 64
          BC113018.1 - 6 - 7 1087 64
          BC113019.1 - 6 - 7 1084 64
          BC128031.1 - 6 - 7 984 64
          M60315.1 - 5 - 6 1441 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006873.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000062.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BMP6

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          osteoblast differentiation
          eye development
          kidney development
          positive regulation of endothelial cell proliferation
          endochondral ossification
          type B pancreatic cell development
          cellular iron ion homeostasis
          inflammatory response
          immune response
          positive regulation of pathway-restricted SMAD protein phosphorylation
          response to activity
          positive regulation of bone mineralization
          BMP signaling pathway
          male genitalia development
          positive regulation of lipopolysaccharide-mediated signaling pathway
          response to magnesium ion
          positive regulation of chondrocyte differentiation
          positive regulation of aldosterone biosynthetic process
          response to retinoic acid
          growth
          regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of endothelial cell differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of epithelial cell proliferation
          positive regulation of protein secretion
          cartilage development
          response to glucocorticoid
          positive regulation of SMAD protein import into nucleus
          SMAD protein signal transduction
          multicellular organismal iron ion homeostasis
          cellular response to mechanical stimulus
          cellular response to iron ion
          cellular response to BMP stimulus
          positive regulation of DNA-dependent DNA replication
          positive regulation of aldosterone secretion
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity
          protein heterodimerization activity
          BMP receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000609.hg.1 TC06000062.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000609.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000609.hg.1 ENST00000537240.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000609.hg.1 ENST00000540959.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000609.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147084.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147084.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147084.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147084.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600147084.hg.1 TC0600006873.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 ENST00000283147.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006873.hg.1 NM_001718.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0