Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01100182_m1
          See other CACNG8 GE Assays ›
          Gene Symbol:
          CACNG8
          Gene Name:
          calcium voltage-gated channel auxiliary subunit gamma 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 53962937 - 53990215 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01100182_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CACNG8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031895.6 NP_114101.4 2 - 3 576 73
          GenBank mRNA AF234892.1 - 2 - 3 391 73
          AF288388.1 - 2 - 3 337 73
          AF361354.1 - 2 - 3 478 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008827.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000869.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CACNG8

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          calcium ion transport
          transmission of nerve impulse
          ion transmembrane transport
          positive regulation of synaptic transmission, glutamatergic
          calcium ion transmembrane transport
          postsynaptic neurotransmitter receptor diffusion trapping
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          voltage-gated calcium channel activity
          calcium channel regulator activity
          calcium channel activity
          channel regulator activity
          protein phosphatase 2B binding
          ionotropic glutamate receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19011633.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19011633.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19011633.hg.1 TC19000869.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 ENST00000401179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008827.hg.1 NR_030632.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900168721.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168721.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900168721.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168721.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900168721.hg.1 TC1900008827.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000270458.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 ENST00000401179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NM_031895.5 Refseq
          Affymetrix Human Clariom D Assay TC1900008827.hg.1 NR_030632.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline