| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761063 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761072 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326525 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326535 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761054 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326515 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761066 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761052 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326538 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326533 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326543 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761060 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326527 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761073 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761061 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761070 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326522 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761069 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761068 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761047 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761071 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08326529 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761057 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761051 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28761050 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
34094 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
716669 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
AB191261.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
AB209840.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
AJ276395.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
AJ535086.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
AL832202.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
AL832771.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BC078656.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BC100030.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BC117176.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX537590.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX538017.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX538018.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX538045.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX640608.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX640731.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX640802.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX640803.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX640875.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX640999.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
BX649182.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
CR749281.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
CR749316.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_002026.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_054034.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_212474.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_212475.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_212476.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_212478.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
NM_212482.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
136386 |
X02761.1 |
genbank_mrna_human |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P661641 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P661641 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
26299 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
1152 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
36159 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P11432354 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P661641 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_24_P661641 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326601 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326573 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761116 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326560 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761105 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761120 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761100 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761127 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761094 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761106 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761096 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16554247 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761138 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761118 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326561 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761087 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761134 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761098 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326598 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761112 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761082 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761086 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761142 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761110 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761109 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761137 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761097 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761103 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326599 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761083 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326550 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761139 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761092 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761122 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761115 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761099 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326578 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761089 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761140 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326555 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761113 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761101 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761123 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761083 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761140 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761092 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761123 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761098 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761142 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761122 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326599 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761137 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761089 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326601 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761139 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761100 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761094 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761112 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761097 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761087 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761103 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761096 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761099 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761134 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326561 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761101 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326555 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761127 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761082 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761115 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761110 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761120 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326578 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761116 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326560 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761109 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326550 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326598 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761105 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08326573 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761113 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761106 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761086 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761118 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16554247 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28761138 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_24_P661641 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_24_P514517 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P11432354 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P11432354 |
- |
- |