| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09003643.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09003643.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09003643.hg.1 |
TC09000343.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900151396.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900151396.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900151396.hg.1 |
ENST00000346234.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900151396.hg.1 |
NM_012383.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900151396.hg.1 |
TC0900007604.hg.1 |
Clariom D TC |