Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01104475_m1
          See other SLC6A1 GE Assays ›
          Gene Symbol:
          SLC6A1
          Gene Name:
          solute carrier family 6 member 1
          Gene Aliases:
          GABATHG, GABATR, GAT1, MAE, hGAT-1
          Chromosome Location:
          Chr.3: 10992748 - 11039247 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01104475_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC6A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001348250.2 NP_001335179.1 5 - 6 807 64
          NM_001348251.2 NP_001335180.1 5 - 6 617 64
          NM_001348252.2 NP_001335181.1 4 - 5 546 64
          NM_001348253.2 NP_001335182.1 4 - 5 485 64
          NM_003042.4 NP_003033.3 5 - 6 868 64
          XM_005265411.6 XP_005265468.1 5 - 6 3169 64
          XM_011534027.4 XP_011532329.1 5 - 6 3859 64
          XM_017007071.3 XP_016862560.1 6 - 7 2651 64
          XM_017007072.3 XP_016862561.1 6 - 7 1961 64
          GenBank mRNA AK295702.1 - 2 - 3 245 64
          BC033904.1 - 5 - 6 850 64
          BC057291.1 - 5 - 6 862 64
          X54673.1 - 4 - 5 705 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300006599.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000063.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001348252.2, NM_001348253.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC6A1

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          neurotransmitter transport
          amino acid transport
          chemical synaptic transmission
          memory
          associative learning
          sodium ion transmembrane transport
          synapse organization
          gamma-aminobutyric acid reuptake
          gamma-aminobutyric acid import
          inorganic anion import across plasma membrane
          sodium ion import across plasma membrane
          neurotransmitter reuptake
          transport across blood-brain barrier
          chloride transmembrane transport
          sodium:amino acid symporter activity
          gamma-aminobutyric acid:sodium symporter activity
          protein binding
          gamma-aminobutyric acid transmembrane transporter activity
          symporter activity
          sodium:chloride symporter activity
          transmembrane transporter activity
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03001379.hg.1 ENST00000287766.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03001379.hg.1 ENST00000536032.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03001379.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03001379.hg.1 TC03000063.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300145800.hg.1 TC0300006599.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 123232 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline