Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01105608_g1
          See other SLC25A1 GE Assays ›
          Gene Symbol:
          SLC25A1
          Gene Name:
          solute carrier family 25 member 1
          Gene Aliases:
          CTP, D2L2AD, SEA, SLC20A3
          Chromosome Location:
          Chr.22: 19175575 - 19178863 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01105608_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC25A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256534.1 NP_001243463.1 2 - 3 473 75
          NM_001287387.1 NP_001274316.1 3 - 4 327 75
          NM_005984.4 NP_005975.1 3 - 4 492 75
          GenBank mRNA AK292313.1 - 3 - 4 396 75
          AK298204.1 - 3 - 4 325 75
          BC004980.1 - 3 - 4 353 2957917 75
          BC008061.2 - 3 - 4 355 2958945 75
          BC018590.1 - 3 - 4 250 75
          BG473148.1 - 2 - 3 447 75
          BP265446.1 - 3 - 4 327 75
          DR006840.1 - 3 - 4 394 75
          HM037273.1 - 2 - 3 328 75
          L75823.1 - 3 - 4 401 75
          L77567.1 - 3 - 4 374 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007976.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000494.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK298204.1, NM_001287387.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC25A1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          gluconeogenesis
          translation
          citrate transport
          long-chain fatty-acyl-CoA biosynthetic process
          tricarboxylic acid transmembrane transport
          structural constituent of ribosome
          citrate transmembrane transporter activity
          tricarboxylic acid transmembrane transporter activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700007750.hg.1 ENST00000395415.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008654.hg.1 ENST00000528351.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000215882.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000451283.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000461267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000468824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000468824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000468824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000468824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 ENST00000470922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001256534.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_001287387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NM_005984.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007976.hg.1 NR_046298.2 Refseq
          Affymetrix Human Clariom D Assay TC0700007750.hg.1 ENST00000395415.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008654.hg.1 ENST00000528351.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 150170 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline