| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013029.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013029.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013029.hg.1 |
ENST00000536181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013029.hg.1 |
ENST00000538942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013029.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X013029.hg.1 |
TC0X000978.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000468431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009486.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
ENST00000378069.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
ENST00000487544.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
NM_000898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164811.hg.1 |
TC0X00009486.hg.1 |
Clariom D TC |