| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01016531.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01016531.hg.1 |
ENST00000369409.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01016531.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01016531.hg.1 |
ENST00000537497.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01016531.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01016531.hg.1 |
TC01001043.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000462324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000469443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000493622.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
ENST00000496756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009621.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
ENST00000369407.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
ENST00000369409.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
ENST00000482968.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
NM_006623.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100169269.hg.1 |
TC0100009621.hg.1 |
Clariom D TC |