| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000630332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
ENST00000630332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006039.hg.1 |
TC03000296.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006039.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006039.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006040.hg.1 |
TC03000296.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006040.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006040.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300152447.hg.1 |
TC0300007385.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000630332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
ENST00000630332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007385.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
ENST00000296452.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
ENST00000467456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
NM_003458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300152446.hg.1 |
TC0300007385.hg.1 |
Clariom D TC |