Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01110766_m1
          See other GLI1 GE Assays ›
          Gene Symbol:
          GLI1
          Gene Name:
          GLI family zinc finger 1
          Gene Aliases:
          GLI, PAPA8, PPD1
          Chromosome Location:
          Chr.12: 57459785 - 57472268 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01110766_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GLI1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160045.2 NP_001153517.1 8 - 9 1533 83
          NM_001167609.2 NP_001161081.1 9 - 10 1630 83
          NM_005269.3 NP_005260.1 10 - 11 1753 83
          XM_011538189.3 XP_011536491.1 10 - 11 1615 83
          GenBank mRNA BC013000.2 - 9 - 10 1347 83
          GQ890670.2 - 8 - 9 1186 83
          X07384.1 - 10 - 11 1387 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007882.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000530.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GLI1

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          smoothened signaling pathway
          spermatogenesis
          spermatid development
          ventral midline development
          positive regulation of cell proliferation
          regulation of smoothened signaling pathway
          response to wounding
          anatomical structure morphogenesis
          epidermal cell differentiation
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          regulation of gene expression
          cerebellar cortex morphogenesis
          pituitary gland development
          cell differentiation
          lung development
          positive regulation of cell migration
          prostate gland development
          regulation of osteoblast differentiation
          positive regulation of DNA replication
          positive regulation of smoothened signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract morphogenesis
          notochord regression
          positive regulation of cardiac muscle cell proliferation
          negative regulation of canonical Wnt signaling pathway
          liver regeneration
          positive regulation of cell cycle G1/S phase transition
          regulation of hepatocyte proliferation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          protein binding
          microtubule binding
          zinc ion binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000228682.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000527742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000527742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000527742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000527742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000528467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000530789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000530789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000530789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000530789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000530789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000532291.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000543426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 ENST00000546141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001160045.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_001167609.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007882.hg.1 NM_005269.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline