Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01118900_m1
          See other ACVR2B GE Assays ›
          Gene Symbol:
          ACVR2B
          Gene Name:
          activin A receptor type 2B
          Gene Aliases:
          ACTRIIB, ActR-IIB, HTX4
          Chromosome Location:
          Chr.3: 38454299 - 38493142 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01118900_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACVR2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106.3 NP_001097.2 1 - 2 74 72
          GenBank mRNA BC096243.1 - 1 - 2 54 40005760 72
          BC096244.1 - 1 - 2 54 40005761 72
          BC096245.1 - 1 - 2 54 40005763 72
          BC099642.4 - 1 - 2 54 40005762 72
          BP352716.1 - 1 - 2 37 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300007077.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000192.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC096245.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ACVR2B

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          gastrulation with mouth forming second
          kidney development
          lymphangiogenesis
          blood vessel remodeling
          regulation of transcription, DNA-templated
          signal transduction
          transmembrane receptor protein serine/threonine kinase signaling pathway
          determination of left/right symmetry
          mesoderm development
          heart development
          response to glucose
          post-embryonic development
          anterior/posterior pattern specification
          signal transduction by protein phosphorylation
          insulin secretion
          lung development
          positive regulation of bone mineralization
          BMP signaling pathway
          pancreas development
          activation of protein kinase activity
          activin receptor signaling pathway
          positive regulation of activin receptor signaling pathway
          organ growth
          odontogenesis of dentin-containing tooth
          positive regulation of osteoblast differentiation
          embryonic foregut morphogenesis
          skeletal system morphogenesis
          palate development
          lymphatic endothelial cell differentiation
          artery development
          venous blood vessel development
          retina vasculature development in camera-type eye
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          activin receptor activity, type II
          growth factor binding
          inhibin binding
          metal ion binding
          activin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03003739.hg.1 ENST00000352511.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03003739.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03003739.hg.1 TC03000192.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03003737.hg.1 ENST00000352511.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03003737.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03003737.hg.1 TC03000192.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149548.hg.1 TC0300007077.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007077.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 ENST00000352511.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 ENST00000461232.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 ENST00000465020.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 NM_001106.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149551.hg.1 TC0300007077.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline