| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007518.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007518.hg.1 |
ENST00000323108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007518.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007518.hg.1 |
TC06004086.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198822.hg.1 |
TC0600014134.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000288390.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000323108.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000506468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
ENST00000515220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014134.hg.1 |
NM_145026.3 |
Refseq |