| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050178.hg.1 |
ENST00000359650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050178.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050178.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050178.hg.1 |
TC01003259.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219078.hg.1 |
TC0100015855.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000359650.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000368739.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
ENST00000490266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015855.hg.1 |
NM_020393.3 |
Refseq |