Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01123453_g1
          See other PYGO2 GE Assays ›
          Gene Symbol:
          PYGO2
          Gene Name:
          pygopus family PHD finger 2
          Gene Aliases:
          1190004M21Rik
          Chromosome Location:
          Chr.1: 154957026 - 154961782 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01123453_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PYGO2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138300.3 NP_612157.1 1 - 2 310 60
          GenBank mRNA AF289598.1 - 1 - 2 62 60
          AF457208.1 - 1 - 2 104 60
          AK092389.1 - 1 - 2 259 60
          BC006132.1 - 1 - 2 276 3627860 60
          BC032099.2 - 1 - 2 276 4126360 60
          CN260857.1 - 1 - 2 313 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018496.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01006380.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF289598.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PYGO2

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          kidney development
          lens development in camera-type eye
          spermatid nucleus differentiation
          brain development
          post-embryonic development
          mammary gland development
          regulation of mammary gland epithelial cell proliferation
          regulation of histone acetylation
          positive regulation of chromatin binding
          developmental growth
          regulation of histone H3-K4 methylation
          palate development
          canonical Wnt signaling pathway
          beta-catenin-TCF complex assembly
          chromatin binding
          protein binding
          zinc ion binding
          histone acetyltransferase regulator activity
          histone binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01050948.hg.1 TC01006380.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050948.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050948.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050949.hg.1 TC01006380.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050949.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050947.hg.1 TC01006380.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050947.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050947.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050947.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243235.hg.1 TC0100018496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100243236.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243236.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243236.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243236.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243236.hg.1 TC0100018496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368456.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018496.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243237.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243237.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243237.hg.1 ENST00000368457.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243237.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243237.hg.1 TC0100018496.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100243238.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243238.hg.1 NM_138300.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243238.hg.1 TC0100018496.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 149740 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline