Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01123677_m1
          See other A3GALT2 GE Assays ›
          Gene Symbol:
          A3GALT2
          Gene Name:
          alpha 1,3-galactosyltransferase 2
          Gene Aliases:
          A3GALT2P, IGB3S, IGBS3S
          Chromosome Location:
          Chr.1: 33306766 - 33321098 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01123677_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A3GALT2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080438.1 NP_001073907.1 4 - 5 331 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013650.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          lipid metabolic process
          glycosphingolipid biosynthetic process
          lipid glycosylation
          alpha-1,3-galactosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring hexosyl groups
          metal ion binding
          N-acetyllactosaminide 3-alpha-galactosyltransferase activity

          MicroArray Probes:

          Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000625117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000625117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000628643.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000628643.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630909.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007720.hg.1 ENST00000630909.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000457957.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000587696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000588828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000624453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000625117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000625117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627130.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000627421.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000628643.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000628643.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630229.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630909.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007720.hg.1 ENST00000630909.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100201537.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100201537.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100201537.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100201537.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100201537.hg.1 TC0100013650.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 ENST00000442999.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 NM_001080438.1 Refseq
          Affymetrix Human Clariom D Assay TC0100013650.hg.1 NM_001080438.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 128702 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1668111 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2295559 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1668111 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1668111 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2295559 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1668111 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1668111 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2295559 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1668111 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2295559 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1668111 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2295559 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline