Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01123934_g1
          See other RDH5 GE Assays ›
          Gene Symbol:
          RDH5 ABO
          Gene Name:
          retinol dehydrogenase 5
          ABO, alpha 1-3-N-acetylgalactosaminyltransferase and alpha 1-3-galactosyltransferase
          Gene Aliases:
          9cRDH, HSD17B9, RDH1, SDR9C5
          A3GALNT, A3GALT1, GTB, NAGAT
          Chromosome Location:
          Chr.12: 55720393 - 55724705 on Build GRCh38
          Chromosome Location:
          Chr.9: 76358 - 101295 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01123934_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RDH5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199771.3 NP_001186700.1 2 - 3 465 69
          NM_002905.5 NP_002896.2 2 - 3 460 69
          GenBank mRNA AK293313.1 - 2 - 3 442 69
          AK307157.1 - 2 - 3 313 69
          BC028298.1 - 2 - 3 445 69
          BI765016.1 - 2 - 3 397 69
          BP381700.1 - 2 - 3 486 69
          U43559.1 - 2 - 3 379 69
          U89717.1 - 2 - 3 416 69
          Gene Symbol: ABO
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA PV266526.1 - 2 - 3 379 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012641.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12003299.hg.1, TC12003300.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RDH5
          Click here to find siRNAs for Gene ABO

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          lipid metabolic process
          visual perception
          steroid metabolic process
          retinol metabolic process
          retinol dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          protein homodimerization activity
          androsterone dehydrogenase activity
          androstan-3-alpha,17-beta-diol dehydrogenase activity
          11-cis-retinol dehydrogenase (NAD+) activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000257899.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000257899.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000257899.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000257899.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000257899.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000257899.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000549424.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000549424.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000549424.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000550412.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000551946.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000551946.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000551946.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000551946.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000551946.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 ENST00000551946.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012642.hg.1 NR_037658.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000257895.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547072.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000547301.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548123.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548123.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548123.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548123.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548123.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000548486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000550608.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000551444.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000551444.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000551444.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000551444.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000551444.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000552930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000552930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000552930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000552930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000552930.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553187.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553187.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553187.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553187.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553187.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 ENST00000553187.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_001199771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012641.hg.1 NM_002905.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-djsrz:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline