| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002483.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002483.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002483.hg.1 |
TC12000200.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200148584.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148584.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148584.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148584.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148584.hg.1 |
TC1200006951.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
ENST00000266395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006951.hg.1 |
NM_006205.2 |
Refseq |