Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01124217_g1
          See other IRX3 GE Assays ›
          Gene Symbol:
          IRX3
          Gene Name:
          iroquois homeobox 3
          Gene Aliases:
          IRX-1, IRXB1
          Chromosome Location:
          Chr.16: 54283304 - 54286787 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01124217_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IRX3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024336.3 NP_077312.2 1 - 2 1004 100
          XM_005256139.4 XP_005256196.1 1 - 2 1004 100
          GenBank mRNA AY335943.1 - 1 - 2 683 100
          BC023667.2 - 1 - 2 650 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010363.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16001124.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IRX3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          metanephros development
          Purkinje myocyte development
          atrioventricular bundle cell differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          mesoderm development
          neuron differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cell development
          His-Purkinje system cell differentiation
          proximal/distal pattern formation involved in nephron development
          specification of loop of Henle identity
          energy homeostasis
          regulation of cell communication by electrical coupling involved in cardiac conduction
          positive regulation of gap junction assembly
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16015641.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015641.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015641.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015641.hg.1 TC16001124.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16015642.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015642.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015642.hg.1 TC16001124.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16015643.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015643.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015643.hg.1 TC16001124.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015640.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015640.hg.1 NM_024336.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015640.hg.1 TC16001124.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1600179506.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179506.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179506.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179506.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179506.hg.1 TC1600010363.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179503.hg.1 TC1600010363.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179505.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179505.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179505.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179505.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179505.hg.1 TC1600010363.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179504.hg.1 TC1600010363.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558054.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 ENST00000558180.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010363.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179507.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179507.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179507.hg.1 ENST00000329734.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1600179507.hg.1 NM_024336.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600179507.hg.1 TC1600010363.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline