| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000261197.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000381424.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000381424.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000381424.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000381424.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000381424.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000390870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000396037.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000404299.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000508924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000508924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000508924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000508924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000508924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000538256.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000538256.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000540824.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000540824.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000540824.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000540824.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000540824.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000540824.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000541868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000541868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000541868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000541868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000541868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000541868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
ENST00000545494.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_001304450.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012775.hg.1 |
NM_003034.3 |
Refseq |