Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01127791_m1
          See other MINPP1 GE Assays ›
          Gene Symbol:
          MINPP1
          Gene Name:
          multiple inositol-polyphosphate phosphatase 1
          Gene Aliases:
          HIPER1, MINPP2, MIPP
          Chromosome Location:
          Chr.10: 87502792 - 87553461 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01127791_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MINPP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001178118.1 NP_001171589.1 2 - 3 609 119
          NM_004897.4 NP_004888.2 2 - 3 1290 119
          XM_006718078.2 XP_006718141.1 2 - 3 2964 119
          XM_011540379.2 XP_011538681.1 3 - 4 976 119
          XM_017016965.1 XP_016872454.1 2 - 3 2964 119
          XM_017016966.1 XP_016872455.1 3 - 4 995 119
          GenBank mRNA AF046914.1 - 2 - 3 840 119
          AF084943.1 - 2 - 3 860 119
          AK304624.1 - 3 - 4 481 119
          AK309176.1 - 2 - 3 608 119
          AK312937.1 - 2 - 3 881 119
          AL050356.1 - 2 - 3 863 119
          AY358938.1 - 2 - 3 845 119
          BC032504.1 - 2 - 3 842 5243440 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008351.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000619.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MINPP1

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          polyphosphate metabolic process
          dephosphorylation
          bone mineralization
          inositol phosphate metabolic process
          acid phosphatase activity
          phosphohistidine phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
          bisphosphoglycerate 3-phosphatase activity
          inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
          inositol hexakisphosphate 2-phosphatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10006785.hg.1 ENST00000371996.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006785.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006785.hg.1 ENST00000546140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006785.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006785.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006785.hg.1 TC10000619.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006783.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006783.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006783.hg.1 TC10000619.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000156558.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156558.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156558.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156558.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156558.hg.1 TC1000008351.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156559.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156559.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156559.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156559.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156559.hg.1 TC1000008351.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156561.hg.1 TC1000008351.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline