Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01128552_m1
          See other WARS2 GE Assays ›
          Gene Symbol:
          WARS2
          Gene Name:
          tryptophanyl tRNA synthetase 2, mitochondrial
          Gene Aliases:
          NEMMLAS, PKDYS3, TrpRS, mtTrpRS
          Chromosome Location:
          Chr.1: 119031216 - 119140672 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01128552_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WARS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001378226.1 NP_001365155.1 3 - 4 438 90
          NM_001378227.1 NP_001365156.1 4 - 5 629 90
          NM_001378230.1 NP_001365159.1 3 - 4 544 90
          NM_001378231.1 NP_001365160.1 2 - 3 353 90
          NM_015836.4 NP_056651.1 2 - 3 353 90
          NM_201263.2 NP_957715.1 2 - 3 372 90
          XM_005270350.4 XP_005270407.1 2 - 3 462 90
          XM_017000038.2 XP_016855527.1 2 - 3 372 90
          XM_024449826.2 XP_024305594.1 3 - 4 725 90
          XM_024449860.2 XP_024305628.1 2 - 3 640 90
          GenBank mRNA AJ242739.1 - 2 - 3 353 90
          AK223197.1 - 2 - 3 350 90
          AK296088.1 - 2 - 3 337 90
          AK296799.1 - 2 - 3 353 90
          AK299547.1 - 2 - 3 311 90
          AK313740.1 - 2 - 3 370 90
          BC021722.1 - 2 - 3 351 90
          BC039889.1 - 2 - 3 348 90
          BC044575.1 - 2 - 3 355 90
          BM834499.1 - 2 - 3 364 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018471.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003044.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene WARS2

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          translation
          tRNA aminoacylation for protein translation
          tryptophanyl-tRNA aminoacylation
          positive regulation of angiogenesis
          mitochondrial tryptophanyl-tRNA aminoacylation
          nucleotide binding
          aminoacyl-tRNA ligase activity
          tryptophan-tRNA ligase activity
          ATP binding
          ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 ENST00000497761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01047453.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01047453.hg.1 ENST00000369426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01047453.hg.1 ENST00000537870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01047453.hg.1 NM_015836.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01047453.hg.1 NM_201263.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01047453.hg.1 TC01003044.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100242712.hg.1 TC0100018471.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000235521.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000369426.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000495746.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497402.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 ENST00000497761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_015836.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq
          Affymetrix Human Clariom D Assay TC0100018471.hg.1 NM_201263.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline