Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01368705_m1
          See other LOC284898 GE Assays ›
          Gene Symbol:
          LOC284898
          Gene Name:
          uncharacterized LOC284898
          Gene Aliases:
          -
          Chromosome Location:
          Chr.22: 27048144 - 27060518 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01368705_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC284898
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318329.1 NP_001305258.1 1 - 2 410 95
          GenBank mRNA AK055980.1 - 1 - 2 190 95
          BC036876.1 - 1 - 2 397 95
          DA498162.1 - 1 - 2 410 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136318 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136328 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136398 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17272093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455906 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17272087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136396 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136464 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17272082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136385 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136389 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17272089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136336 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136394 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32136326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11456038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11455949 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 148241 AF161482.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 924328 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 277069 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 477410 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 677917 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3825454 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 44322 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3825454 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03609963 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 103330 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P22639448 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03609963 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21299113 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03609954 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13978649 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03609950 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03609966 - -
          Agilent Human 1B Microarray G4111A A_32_P896115 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03609954 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P22639448 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03609963 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P13978649 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562030_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200008329.hg.1 ENST00000453934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008329.hg.1 ENST00000453934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008329.hg.1 ENST00000453934.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008329.hg.1 ENST00000453934.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1562030_at - -
          Human Genome U133 Plus 2.0 Array 1562030_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562030_at5 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 137948 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline