| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000577271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000577271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000577271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700154444.hg.1 |
TC1700007565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000321639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000577271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000577271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000577271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
ENST00000631683.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007565.hg.1 |
NM_001304438.1 |
Refseq |